COMH93D62BD5 Combi-Blocks, Inc.

1-((2-(Trimethylsilyl)ethoxy)carbonyl)azetidine-3-carboxylic acid

  • MDL number MFCD32198809
  • Molecular Weight : 245.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-8582

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-8582
Availability details for
Quantity
  • Availability
  • Add to Cart