COMH93D62C1E Combi-Blocks, Inc.

T-Boc-n-amido-peg3-nhs ester

  • MDL number MFCD31811545
  • Cas Number : 2250216-93-2
  • Molecular Weight : 418.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-8670

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-8670
Availability details for
Quantity
  • Availability
  • Add to Cart