COMH93D62C34 Combi-Blocks, Inc.

Benzenemethanamine, 4-[6-(2-pyridinyl)-1,2,4,5-tetrazin-3-yl]-, tfa salt

  • MDL number MFCD31811498
  • Cas Number : 1345955-29-4
  • Molecular Weight : 264.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-8693

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-8693
Availability details for
Quantity
  • Availability
  • Add to Cart