COMH93D62C37 Combi-Blocks, Inc.

Lithospermic acid

  • MDL number MFCD03427311
  • Cas Number : 28831-65-4
  • Molecular Weight : 538.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-8696

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-8696
Availability details for
Quantity
  • Availability
  • Add to Cart