COMH93D62C4F Combi-Blocks, Inc.

4-Bromo-1-(2,2-difluoroethyl)-3-iodo-1h-pyrrolo[2,3-b]pyridine

  • MDL number MFCD31924957
  • Cas Number : 2358751-17-2
  • Molecular Weight : 386.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-8725

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-8725
Availability details for
Quantity
  • Availability
  • Add to Cart