COMH93D62D4C Combi-Blocks, Inc.

2-(Azetidin-3-yl)-2-methylpropanoic acid tfa

  • MDL number MFCD31705191
  • Cas Number : 2225878-90-8
  • Molecular Weight : 257.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9034

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9034
Availability details for
Quantity
  • Availability
  • Add to Cart