COMH93D62E85 Combi-Blocks, Inc.

3-(4-Bromo-3-methoxyphenoxy)azetidine

  • MDL number MFCD31735227
  • Cas Number : 2229476-74-6
  • Molecular Weight : 258.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9403

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9403
Availability details for
Quantity
  • Availability
  • Add to Cart