COMH93D62E8D Combi-Blocks, Inc.

4-Carboxymethoxy-3-methoxy-benzoic acid methyl ester

  • MDL number MFCD31716707
  • Cas Number : 95046-98-3
  • Molecular Weight : 240.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9412

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9412
Availability details for
Quantity
  • Availability
  • Add to Cart