COMH93D62EA2 Combi-Blocks, Inc.

1-Methyl-3-(trifluoromethyl)-1h-pyrazole-5-carbohydrazide

  • MDL number MFCD03419674
  • Cas Number : 1001518-78-0
  • Molecular Weight : 208.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9442

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9442
Availability details for
Quantity
  • Availability
  • Add to Cart