COMH93D62EAB Combi-Blocks, Inc.

4-((4-Bromo-3-nitrophenyl)sulfonyl)morpholine

  • MDL number MFCD31760185
  • Cas Number : 1604059-85-9
  • Molecular Weight : 351.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9451

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9451
Availability details for
Quantity
  • Availability
  • Add to Cart