COMH93D62EAC Combi-Blocks, Inc.

Methyl 1-[6-(trifluoromethyl)-3-pyridyl]cyclopropanecarboxylate

  • MDL number MFCD31729393
  • Cas Number : 2119721-57-0
  • Molecular Weight : 245.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9452

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9452
Availability details for
Quantity
  • Availability
  • Add to Cart