COMH93D62EB7 Combi-Blocks, Inc.

3-[4-Chloro-3-(trifluoromethyl)-1h-pyrazol-1-yl]propanoic acid

  • MDL number MFCD04969243
  • Cas Number : 1006470-23-0
  • Molecular Weight : 242.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9466

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9466
Availability details for
Quantity
  • Availability
  • Add to Cart