COMH93D62ECA Combi-Blocks, Inc.

1-(5-Pent-4-enyloxy-pyridin-2-yl)-piperazine

  • MDL number MFCD31700238
  • Cas Number : 2270912-48-4
  • Molecular Weight : 247.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9487

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9487
Availability details for
Quantity
  • Availability
  • Add to Cart