COMH93D62EF5 Combi-Blocks, Inc.

2-Bromo-4-isobutoxy-1-(trifluoromethyl)benzene

  • MDL number MFCD32198870
  • Molecular Weight : 297.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9540

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9540
Availability details for
Quantity
  • Availability
  • Add to Cart