COMH93D62FC0 Combi-Blocks, Inc.

[1-(4-Thiophen-2-yl-phenyl)-cyclopropyl]-methanol

  • MDL number MFCD31689197
  • Cas Number : 2270912-69-9
  • Molecular Weight : 230.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9792

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9792
Availability details for
Quantity
  • Availability
  • Add to Cart