COMH93D63002 Combi-Blocks, Inc.

1-(2,3,5,6-Tetrafluorobenzyl)-1h-pyrazol-4-amine

  • MDL number MFCD04967238
  • Cas Number : 1005630-54-5
  • Molecular Weight : 245.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9869

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9869
Availability details for
Quantity
  • Availability
  • Add to Cart