COMH93D63021 Combi-Blocks, Inc.

6-Fluoro-3-indoxyl-beta-d-galactopyranoside

  • MDL number MFCD00798396
  • Cas Number : 207727-11-5
  • Molecular Weight : 313.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9905

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9905
Availability details for
Quantity
  • Availability
  • Add to Cart