COMH93D63024 Combi-Blocks, Inc.

Dbco-peg12-nhs ester

  • MDL number MFCD31811532
  • Cas Number : 2093934-94-0
  • Molecular Weight : 1002.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9908

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9908
Availability details for
Quantity
  • Availability
  • Add to Cart