COMH93D63030 Combi-Blocks, Inc.

Methyl (2-chloro-4-ethylphenoxy)acetate

  • MDL number MFCD31693786
  • Cas Number : 2113558-73-7
  • Molecular Weight : 228.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9927

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9927
Availability details for
Quantity
  • Availability
  • Add to Cart