COMH93D63058 Combi-Blocks, Inc.

3-(4-Methanesulfonylphenyl)-2-methylpropanoic acid

  • MDL number MFCD31716477
  • Cas Number : 1505635-18-6
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9976

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HE-9976
Availability details for
Quantity
  • Availability
  • Add to Cart