COMH93D632D1 Combi-Blocks, Inc.

((E)-1-But-1-enyl)-pyrrolidine

  • MDL number MFCD19981423
  • Cas Number : 56176-56-8
  • Molecular Weight : 125.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-0724

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-0724
Availability details for
Quantity
  • Availability
  • Add to Cart