COMH93D6338B Combi-Blocks, Inc.

Ethyl 5-amino-4-bromo-1-(4-fluorophenyl)pyrazole-3-carboxylate

  • MDL number MFCD23135137
  • Cas Number : 1427022-75-0
  • Molecular Weight : 328.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-0952

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-0952
Availability details for
Quantity
  • Availability
  • Add to Cart