COMH93D63400 Combi-Blocks, Inc.

Galnac beta(1-4)glcnac-beta-pnp

  • MDL number MFCD15072176
  • Cas Number : 872578-72-8
  • Molecular Weight : 545.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-1085

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-1085
Availability details for
Quantity
  • Availability
  • Add to Cart