COMH93D6346A Combi-Blocks, Inc.

N-Methyl-n-(4-pyrimidin-5-ylbenzyl)amine

  • MDL number MFCD07772857
  • Cas Number : 859850-85-4
  • Molecular Weight : 199.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-1214

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-1214
Availability details for
Quantity
  • Availability
  • Add to Cart