COMH93D634A9 Combi-Blocks, Inc.

Trans-5-trimethylsilyl-2-penten-4-yn-1-ol

  • MDL number MFCD09039152
  • Cas Number : 97514-97-1
  • Molecular Weight : 154.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-1282

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-1282
Availability details for
Quantity
  • Availability
  • Add to Cart