COMH93D6369B Combi-Blocks, Inc.

Schisanhenol

  • MDL number MFCD28411506
  • Cas Number : 69363-14-0
  • Molecular Weight : 402.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-1859

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-1859
Availability details for
Quantity
  • Availability
  • Add to Cart