COMH93D6380D Combi-Blocks, Inc.

3-[5-Methyl-3-(trifluoromethyl)-1h-pyrazol-1-yl]butanenitrile

  • MDL number MFCD06739386
  • Cas Number : 1006348-45-3
  • Molecular Weight : 217.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-2304

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-2304
Availability details for
Quantity
  • Availability
  • Add to Cart