COMH93D63836 Combi-Blocks, Inc.

4-Bromo-3-methyl-isothiazole-5-carbaldehyde

  • MDL number MFCD27937097
  • Cas Number : 89283-96-5
  • Molecular Weight : 206.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-2354

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-2354
Availability details for
Quantity
  • Availability
  • Add to Cart