COMH93D63866 Combi-Blocks, Inc.

5-(Difluoromethyl)thiophene-2-carbonitrile

  • MDL number MFCD29767715
  • Cas Number : 2112657-85-7
  • Molecular Weight : 159.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-2412

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-2412
Availability details for
Quantity
  • Availability
  • Add to Cart