COMH93D6387D Combi-Blocks, Inc.

5-Bromo-3,4-dimethyl-1h-pyrrole-2-carboxylic acid

  • MDL number MFCD13705413
  • Cas Number : 861034-48-2
  • Molecular Weight : 218.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-2440

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-2440
Availability details for
Quantity
  • Availability
  • Add to Cart