COMH93D63894 Combi-Blocks, Inc.

Glcnac beta(1-3)[glcnac beta(1-6)]galnac-alpha-thr

  • MDL number MFCD15072166
  • Cas Number : 1304646-03-4
  • Molecular Weight : 714.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-2466

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-2466
Availability details for
Quantity
  • Availability
  • Add to Cart