COMH93D638E1 Combi-Blocks, Inc.

3-Azabicyclo[3.1.0]hexan-6-amine dihydrochloride

  • MDL number MFCD08705927
  • Cas Number : 1369494-70-1
  • Molecular Weight : 171.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-2552

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-2552
Availability details for
Quantity
  • Availability
  • Add to Cart