COMH93D63D18 Combi-Blocks, Inc.

3-(2-Iodoethyl)-4-(1h,1h-perfluoroheptyl)pyrrolidine-1-carbonitrile

  • MDL number MFCD22124040
  • Cas Number : 1365808-06-5
  • Molecular Weight : 582.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-3829

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-3829
Availability details for
Quantity
  • Availability
  • Add to Cart