COMH93D63F77 Combi-Blocks, Inc.

4-Chloro-2-(3-chloropyridin-2-yl)-6-methylpyrimidine

  • MDL number MFCD11227227
  • Cas Number : 1089330-86-8
  • Molecular Weight : 240.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-4523

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-4523
Availability details for
Quantity
  • Availability
  • Add to Cart