COMH93D63FEB Combi-Blocks, Inc.

2-Pyrimidinecarbonyl chloride

  • MDL number MFCD18816666
  • Cas Number : 220769-83-5
  • Molecular Weight : 142.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-4660

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-4660
Availability details for
Quantity
  • Availability
  • Add to Cart