COMH93D6412E Combi-Blocks, Inc.

4-(4-Bromophenyl)butan-2-ol

  • MDL number MFCD16150637
  • Cas Number : 223554-16-3
  • Molecular Weight : 229.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-5024

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-5024
Availability details for
Quantity
  • Availability
  • Add to Cart