COMH93D6414E Combi-Blocks, Inc.

N-(4-Trifluoromethyl-phenyl)-guanidine

  • MDL number MFCD10000732
  • Cas Number : 130066-24-9
  • Molecular Weight : 203.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-5061

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-5061
Availability details for
Quantity
  • Availability
  • Add to Cart