COMH93D643E0 Combi-Blocks, Inc.

3,6-Difluoro-2-methoxybenzamide

  • MDL number MFCD18394893
  • Cas Number : 1261439-98-8
  • Molecular Weight : 187.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-5816

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-5816
Availability details for
Quantity
  • Availability
  • Add to Cart