COMH93D644BA Combi-Blocks, Inc.

3-[5-Cyclopropyl-3-(trifluoromethyl)-1h-pyrazol-1-yl]propanenitrile

  • MDL number MFCD08558158
  • Cas Number : 1006334-01-5
  • Molecular Weight : 229.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-6072

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-6072
Availability details for
Quantity
  • Availability
  • Add to Cart