COMH93D6458A Combi-Blocks, Inc.

4,6-Dichloro-5-(difluoromethyl)pyrimidin-2-amine

  • MDL number MFCD29767531
  • Cas Number : 2091217-55-7
  • Molecular Weight : 214

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-6312

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-6312
Availability details for
Quantity
  • Availability
  • Add to Cart