COMH93D64725 Combi-Blocks, Inc.

Ethyl 2-(4-fluorophenyl)-2-hydroxyacetate

  • MDL number MFCD16661866
  • Cas Number : 7550-03-0
  • Molecular Weight : 198.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-6805

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-6805
Availability details for
Quantity
  • Availability
  • Add to Cart