COMH93D647AD Combi-Blocks, Inc.

3-[1'-Ethyl-5-(trifluoromethyl)-1h,1'h-3,4'-bipyrazol-1-yl]propan-1-amine

  • MDL number MFCD08556271
  • Cas Number : 1006336-85-1
  • Molecular Weight : 287.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-6964

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-6964
Availability details for
Quantity
  • Availability
  • Add to Cart