COMH93D64823 Combi-Blocks, Inc.

2,2-Difluoro-1-phenyl-2-(pyridin-3-yl)ethan-1-one

  • MDL number MFCD22377945
  • Cas Number : 1378261-26-7
  • Molecular Weight : 233.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-7102

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-7102
Availability details for
Quantity
  • Availability
  • Add to Cart