COMH93D648B5 Combi-Blocks, Inc.

2-Amino-2-(3-fluorobicyclo[1.1.1]pentan-1-yl)acetic acid hydrochloride

  • MDL number MFCD29060099
  • Cas Number : 1934367-59-5
  • Molecular Weight : 195.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-7262

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-7262
Availability details for
Quantity
  • Availability
  • Add to Cart