COMH93D64969 Combi-Blocks, Inc.

N-Glcnac-biotin

  • MDL number MFCD10567198
  • Cas Number : 1272755-69-7
  • Molecular Weight : 446.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-7472

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-7472
Availability details for
Quantity
  • Availability
  • Add to Cart