COMH93D64C1B Combi-Blocks, Inc.

3-(2-Fluoro-[1,1'-biphenyl]-4-yl)-1h-pyrazole

  • MDL number MFCD18089275
  • Cas Number : 1287217-55-3
  • Molecular Weight : 238.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-8284

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-8284
Availability details for
Quantity
  • Availability
  • Add to Cart