COMH93D64C3F Combi-Blocks, Inc.

Ethyl 5-amino-1-(3-pyridyl)pyrazole-3-carboxylate

  • MDL number MFCD23135157
  • Cas Number : 1427023-96-8
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-8325

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-8325
Availability details for
Quantity
  • Availability
  • Add to Cart