COMH93D64CBA Combi-Blocks, Inc.

3-[1'-Methyl-5-(trifluoromethyl)-1h,1'h-3,4'-bipyrazol-1-yl]propanenitrile

  • MDL number MFCD08556296
  • Cas Number : 1006353-16-7
  • Molecular Weight : 269.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-8475

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-8475
Availability details for
Quantity
  • Availability
  • Add to Cart