COMH93D64CEA Combi-Blocks, Inc.

3,4-Difluoro-benzimidic acid ethyl ester hydrochloride

  • MDL number MFCD12131139
  • Cas Number : 1205638-72-7
  • Molecular Weight : 221.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-8538

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-8538
Availability details for
Quantity
  • Availability
  • Add to Cart