COMH93D64D56 Combi-Blocks, Inc.

Sialyl lewis x-lactose

  • MDL number MFCD20265602
  • Cas Number : 127923-85-7
  • Molecular Weight : 1145.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HF-8654

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HF-8654
Availability details for
Quantity
  • Availability
  • Add to Cart